accessibility menu, dialog, popup

UserName

CAS RN 797755-07-8

IUPAC Name: 2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
Synonyms: [4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetic acid
Molecular Weight (g/mol): 262.11
Molecular Formula: C14H19BO4
InChi Key: FNLWHBHWDXCWHV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CC(O)=O)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results