accessibility menu, dialog, popup

UserName

AlOOOH3CH3CCH3H3CCH3CH3H3CCH3HCH3Li

CAS RN 79172-99-9

IUPAC Name: lithium(1+) tris[(3-ethylpentan-3-yl)oxy]alumanuide
Synonyms: lithium(1+) tris[(3-ethylpentan-3-yl)oxy]alumanuide
Molecular Weight (g/mol): 380.52
Molecular Formula: C21H46AlLiO3
InChi Key: FXBRQZDWJRAIEB-UHFFFAOYSA-N
SMILES: [Li+].CCC(CC)(CC)O[AlH-](OC(CC)(CC)CC)OC(CC)(CC)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)