accessibility menu, dialog, popup

UserName

CAS RN 7791-12-0

IUPAC Name: λ¹-thallanylium chloride
Synonyms: thallium(I) chloride
Molecular Weight (g/mol): 239.83
Molecular Formula: ClTl
InChi Key: GBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES: [Cl-].[Tl+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results