CAS RN 7791-07-3
IUPAC Name:
sodium hydrate perchlorate
Synonyms:
sodium hydrate perchlorate
Molecular Weight (g/mol):
140.45
Molecular Formula:
ClH2NaO5
InChi Key:
IXGNPUSUVRTQGW-UHFFFAOYSA-M
SMILES:
O.[Na+].[O-][Cl](=O)(=O)=O
1
–
1
of
1
results
1
Sodium Perchlorate Monohydrate (HPLC), Fisher Chemical™
CAS: 7791-07-3 Molecular Formula: ClH2NaO5 Molecular Weight (g/mol): 140.45 MDL Number: MFCD00149164 InChI Key: IXGNPUSUVRTQGW-UHFFFAOYSA-M Synonym: sodium perchlorate monohydrate,sodium perchlorate hydrate,unii-g4h485lhcw,g4h485lhcw,ccris 8893,perchloric acid, sodium salt, monohydrate,sodium hydrate perchlorate,clo4.na.h2o,acmc-1bii1,sodium perchlorate-monohydrate PubChem CID: 516899 IUPAC Name: sodium;perchlorate;hydrate SMILES: O.[O-]Cl(=O)(=O)=O.[Na+]