accessibility menu, dialog, popup

UserName

CAS RN 7790-48-9

IUPAC Name: tetraiodo-λ⁴-tellane
Synonyms: tellurium tetraiodide
Molecular Weight (g/mol): 635.22
Molecular Formula: I4Te
InChi Key: XCOKHDCPVWVFKS-UHFFFAOYSA-N
SMILES: I[Te](I)(I)I
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results