accessibility menu, dialog, popup

UserName

CAS RN 7786-67-6

IUPAC Name: 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
Synonyms: 5-methyl-2-(prop-1-en-2-yl)cyclohexan-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: ZYTMANIQRDEHIO-UHFFFAOYNA-N
SMILES: CC1CCC(C(O)C1)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results