accessibility menu, dialog, popup

UserName

CAS RN 7785-21-9

IUPAC Name: magnesium(2+) ammonium phosphate
Synonyms: ammonium magnesium phosphate
Molecular Weight (g/mol): 137.31
Molecular Formula: H4MgNO4P
InChi Key: MXZRMHIULZDAKC-UHFFFAOYSA-L
SMILES: [NH4+].[Mg++].[O-]P([O-])([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results