accessibility menu, dialog, popup

UserName

AlIII

CAS RN 7784-23-8

IUPAC Name: aluminium(3+) triiodide
Synonyms: aluminium iodide
Molecular Weight (g/mol): 407.69
Molecular Formula: AlI3
InChi Key: CECABOMBVQNBEC-UHFFFAOYSA-K
SMILES: [Al+3].[I-].[I-].[I-]
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results