accessibility menu, dialog, popup

UserName

AgONO

CAS RN 7783-99-5

IUPAC Name: silver(1+) nitrite
Synonyms: silver nitrite
Molecular Weight (g/mol): 153.87
Molecular Formula: AgNO2
InChi Key: KKKDGYXNGYJJRX-UHFFFAOYSA-M
SMILES: [Ag+].[O-]N=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)