accessibility menu, dialog, popup

UserName

FFSn

CAS RN 7783-47-3

IUPAC Name: λ²-tin(2+) difluoride
Synonyms: stannous fluoride
Molecular Weight (g/mol): 156.71
Molecular Formula: F2Sn
InChi Key: ANOBYBYXJXCGBS-UHFFFAOYSA-L
SMILES: [F-].[F-].[Sn++]
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)