accessibility menu, dialog, popup

UserName

HgOSOOO

CAS RN 7783-35-9

IUPAC Name: mercury(2+) sulfate
Synonyms: mercury(II) sulfate mercury(I) sulfate
Molecular Weight (g/mol): 296.65
Molecular Formula: HgO4S
InChi Key: DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES: [Hg++].[O-]S([O-])(=O)=O
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results