Iodic Acid
- (2)
- (8)
- (1)
- (1)
- (1)
- (3)
- (3)
- (1)
- (3)
- (3)
- (2)
- (2)
- (2)
- (3)
Filtered Search Results
Iodic acid, 99.5%, for analysis
CAS: 7782-68-5 Molecular Formula: ILiO3 Molecular Weight (g/mol): 181.84 MDL Number: MFCD00011348 InChI Key: FZAXZVHFYFGNBX-UHFFFAOYSA-M Synonym: unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min PubChem CID: 24345 ChEBI: CHEBI:24857 IUPAC Name: lithium(1+) iodate SMILES: [Li+].[O-][I](=O)=O
| PubChem CID | 24345 |
|---|---|
| CAS | 7782-68-5 |
| Molecular Weight (g/mol) | 181.84 |
| ChEBI | CHEBI:24857 |
| MDL Number | MFCD00011348 |
| SMILES | [Li+].[O-][I](=O)=O |
| Synonym | unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min |
| IUPAC Name | lithium(1+) iodate |
| InChI Key | FZAXZVHFYFGNBX-UHFFFAOYSA-M |
| Molecular Formula | ILiO3 |
Iodic acid, ACS, 99.5% min
CAS: 7782-68-5 Molecular Formula: HIO3 Molecular Weight (g/mol): 175.91 MDL Number: MFCD00011348 InChI Key: FZAXZVHFYFGNBX-UHFFFAOYSA-M Synonym: unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min PubChem CID: 24345 ChEBI: CHEBI:24857 IUPAC Name: lithium(1+) iodate SMILES: [Li+].[O-][I](=O)=O
| PubChem CID | 24345 |
|---|---|
| CAS | 7782-68-5 |
| Molecular Weight (g/mol) | 175.91 |
| ChEBI | CHEBI:24857 |
| MDL Number | MFCD00011348 |
| SMILES | [Li+].[O-][I](=O)=O |
| Synonym | unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min |
| IUPAC Name | lithium(1+) iodate |
| InChI Key | FZAXZVHFYFGNBX-UHFFFAOYSA-M |
| Molecular Formula | HIO3 |
Iodic acid, 99.6%, ACS reagent
CAS: 7782-68-5 Molecular Formula: ILiO3 Molecular Weight (g/mol): 181.84 MDL Number: MFCD00011348 InChI Key: FZAXZVHFYFGNBX-UHFFFAOYSA-M Synonym: unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min PubChem CID: 24345 ChEBI: CHEBI:24857 IUPAC Name: lithium(1+) iodate SMILES: [Li+].[O-][I](=O)=O
| PubChem CID | 24345 |
|---|---|
| CAS | 7782-68-5 |
| Molecular Weight (g/mol) | 181.84 |
| ChEBI | CHEBI:24857 |
| MDL Number | MFCD00011348 |
| SMILES | [Li+].[O-][I](=O)=O |
| Synonym | unii-6u8j18jsbm,6u8j18jsbm,iodicacid,monoiodic acid,caswell no. 499a,iodsaeure,trioxoiodic acid,iodic v acid,sodium iodate min |
| IUPAC Name | lithium(1+) iodate |
| InChI Key | FZAXZVHFYFGNBX-UHFFFAOYSA-M |
| Molecular Formula | ILiO3 |
Iodic acid, 99%
CAS: 7782-68-5 Molecular Formula: ILiO3 Molecular Weight (g/mol): 181.84 MDL Number: MFCD00011348 InChI Key: FZAXZVHFYFGNBX-UHFFFAOYSA-M PubChem CID: 24345 ChEBI: CHEBI:24857 IUPAC Name: lithium(1+) iodate SMILES: [Li+].[O-][I](=O)=O
| PubChem CID | 24345 |
|---|---|
| CAS | 7782-68-5 |
| Molecular Weight (g/mol) | 181.84 |
| ChEBI | CHEBI:24857 |
| MDL Number | MFCD00011348 |
| SMILES | [Li+].[O-][I](=O)=O |
| IUPAC Name | lithium(1+) iodate |
| InChI Key | FZAXZVHFYFGNBX-UHFFFAOYSA-M |
| Molecular Formula | ILiO3 |