CAS RN 7781-49-9
CAS RN 7781-49-9
IUPAC Name:
(E)-N-[(4-iodophenyl)imino]-N'-[(4-nitrophenyl)amino]benzenecarboximidamide
Synonyms:
(E)-N-[(4-iodophenyl)imino]-N'-[(4-nitrophenyl)amino]benzenecarboximidamide
Molecular Weight (g/mol):
471.26
Molecular Formula:
C19H14IN5O2
InChi Key:
FVFWUNAWQMROIF-NKFFIXGESA-N
SMILES:
[O-][N+](=O)C1=CC=C(N\N=C(\N=NC2=CC=C(I)C=C2)C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
INT Formazan, TCI America™
CAS: 7781-49-9 Molecular Formula: C19H14IN5O2 Molecular Weight (g/mol): 471.26 MDL Number: MFCD00007300 InChI Key: FVFWUNAWQMROIF-NKFFIXGESA-N Synonym: 1-(4-Iodophenyl)-5-(4-nitrophenyl)-3-phenylformazan, Iodonitrotetrazolium Formazan PubChem CID: 5485654 IUPAC Name: (E)-N-[(4-iodophenyl)imino]-N'-[(4-nitrophenyl)amino]benzenecarboximidamide SMILES: [O-][N+](=O)C1=CC=C(N\N=C(\N=NC2=CC=C(I)C=C2)C2=CC=CC=C2)C=C1