accessibility menu, dialog, popup

UserName

CAS RN 7774-74-5

IUPAC Name: thiophene-2-thiol
Synonyms: thiophene-2-thiol
Molecular Weight (g/mol): 116.2
Molecular Formula: C4H4S2
InChi Key: SWEDAZLCYJDAGW-UHFFFAOYSA-N
SMILES: SC1=CC=CS1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)