accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 773-76-2

OHClClN

CAS RN 773-76-2

IUPAC Name: 5,7-dichloroquinolin-8-ol
Synonyms: chloroxine chloroxyquinoline quinolor chlorquinol chlorohydroxyquinoline chlofucid quesyl dichlorohydroxyquinoline dichloroxin 哈喹诺
Molecular Weight (g/mol): 214.05
Molecular Formula: C9H5Cl2NO
InChi Key: WDFKMLRRRCGAKS-UHFFFAOYSA-N
SMILES: OC1=C2N=CC=CC2=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.