accessibility menu, dialog, popup

UserName

CAS RN 7712-05-2

IUPAC Name: 4H,6H-thieno[2,3-c]thiophene-2-carboxylic acid
Synonyms: 4H,6H-thieno[2,3-c]thiophene-2-carboxylic acid
Molecular Weight (g/mol): 186.24
Molecular Formula: C7H6O2S2
InChi Key: RBINIIIGSUWMLQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=C(CSC2)S1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results