accessibility menu, dialog, popup

UserName

HHZr

CAS RN 7704-99-6

IUPAC Name: λ²-zirconium(2+) dihydride
Synonyms: λ²-zirconium(2+) dihydride
Molecular Weight (g/mol): 93.24
Molecular Formula: H2Zr
InChi Key: IUJYJRCMJZIAJY-UHFFFAOYSA-N
SMILES: [H-].[H-].[Zr++]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results