CAS RN 7681-65-4
IUPAC Name:
λ¹-copper(1+) iodide
Synonyms:
cuprous iodide
Molecular Weight (g/mol):
190.45
Molecular Formula:
CuI
InChi Key:
LSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES:
[Cu+].[I-]
1
–
3
of
3
results
1
Copper(I) iodide, 98%
CAS: 7681-65-4 MDL Number: MFCD00010978 Synonym: cuprous iodide,copper l iodide,copper i iodide metals basis,marshite,copper i iodide 250g
2
Copper(I) iodide, 98%
CAS: 7681-65-4 Molecular Formula: CuI Molecular Weight (g/mol): 190.45 MDL Number: MFCD00010978 Synonym: cuprous iodide,copper l iodide,copper i iodide metals basis,marshite,copper i iodide 250g
3
Copper(I) iodide, 99.995%, (trace metal basis)
CAS: 7681-65-4 Molecular Formula: CuI Molecular Weight (g/mol): 190.45 MDL Number: MFCD00010978 Synonym: cuprous iodide,copper l iodide,copper i iodide metals basis,marshite,copper i iodide 250g