CAS RN 7661-32-7
IUPAC Name:
1-(4-bromophenyl)pyrrolidin-2-one
Synonyms:
1-(4-bromophenyl)pyrrolidin-2-one
Molecular Weight (g/mol):
240.1
Molecular Formula:
C10H10BrNO
InChi Key:
YINFEFUSAQRZGG-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N1CCCC1=O
1
–
1
of
1
results
1
Promotions Available
1-(4-Bromophenyl)pyrrolidin-2-one, 98%
1-(4-Bromophenyl)pyrrolidin-2-one, 98%
CAS: 7661-32-7 Molecular Formula: C10H10BrNO Molecular Weight (g/mol): 240.1 MDL Number: MFCD00138502 InChI Key: YINFEFUSAQRZGG-UHFFFAOYSA-N Synonym: 1-4-bromophenyl pyrrolidin-2-one,1-4-bromophenyl-2-pyrrolidinone,2-pyrrolidinone, 1-4-bromophenyl,1-4-bromophenyl-2-pyrrolidone,pubchem9356,p-bromo-phenylpyrrolidone,acmc-209p4u,buttpark 32\07-61,n-4-bromophenyl-2-pyrrolidinone,1-4-bromophenyl pyrrolidine-2-one PubChem CID: 736110 IUPAC Name: 1-(4-bromophenyl)pyrrolidin-2-one SMILES: C1CC(=O)N(C1)C2=CC=C(C=C2)Br