accessibility menu, dialog, popup

UserName

Explore New Spotlight Deals on Equipment, Instruments, and More

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 766-93-8

ONH

CAS RN 766-93-8

IUPAC Name: N-cyclohexylformamide
Synonyms: cyclohexylformamide N-zyklohexylformamid
Molecular Weight (g/mol): 127.19
Molecular Formula: C7H13NO
InChi Key: SWGXDLRCJNEEGZ-UHFFFAOYSA-N
SMILES: O=CNC1CCCCC1
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.