accessibility menu, dialog, popup

UserName

H3CCH

CAS RN 765-13-9

IUPAC Name: pentadec-1-yne
Synonyms: 1-pentadecyne
Molecular Weight (g/mol): 208.39
Molecular Formula: C15H28
InChi Key: DONJGKADZJEXRJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results