accessibility menu, dialog, popup

UserName

CAS RN 76420-72-9

IUPAC Name: 2-{2-[(1-carboxy-3-phenylpropyl)amino]propanoyl}pyrrolidine-1-carboxylic acid dihydrate
Synonyms: enalaprilat
Molecular Weight (g/mol): 384.43
Molecular Formula: C18H28N2O7
InChi Key: LRBNLIUVJIKAQR-UHFFFAOYNA-N
SMILES: O.O.CC(NC(CCC1=CC=CC=C1)C(O)=O)C(=O)C1CCCN1C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)