accessibility menu, dialog, popup

UserName

CH3H3C

CAS RN 764-73-8

IUPAC Name: octa-2,6-diyne
Synonyms: octa-2,6-diyne
Molecular Weight (g/mol): 106.17
Molecular Formula: C8H10
InChi Key: PHNFSGFUJPBCLS-UHFFFAOYSA-N
SMILES: CC#CCCC#CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results