CAS RN 76172-73-1
IUPAC Name:
bis(2-phenoxyacetic acid) dihydrate magnesium
Synonyms:
bis(phenoxyacetic acid) dihydrate magnesium
Molecular Weight (g/mol):
364.63
Molecular Formula:
C16H20MgO8
InChi Key:
ZFPUMEQWQQBGMQ-UHFFFAOYSA-N
SMILES:
O.O.[Mg].OC(=O)COC1=CC=CC=C1.OC(=O)COC1=CC=CC=C1
1
–
1
of
1
results
1
Magnesium Phenoxyacetate Dihydrate 98.0+%, TCI America™
CAS: 76172-73-1 Molecular Formula: C16H20MgO8 Molecular Weight (g/mol): 364.633 MDL Number: MFCD00059853 InChI Key: ZFPUMEQWQQBGMQ-UHFFFAOYSA-N Synonym: Phenoxyacetic Acid Magnesium Salt PubChem CID: 131884797 IUPAC Name: magnesium;2-phenoxyacetic acid;dihydrate SMILES: C1=CC=C(C=C1)OCC(=O)O.C1=CC=C(C=C1)OCC(=O)O.O.O.[Mg]