accessibility menu, dialog, popup

UserName

CAS RN 75763-51-8

IUPAC Name: 9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 9-[(2R,3R,4S,5R)-3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol): 267.25
Molecular Formula: C10H13N5O4
InChi Key: PPLSURAOXNSSRH-GPGUAWMRNA-N
SMILES: N[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1N1C=NC2=C1N=CNC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results