CAS RN 75747-14-7
CAS RN 75747-14-7
IUPAC Name:
(4Z,6E,8R,9R,10E,12R,13S,14R,16R)-13-hydroxy-8,14-dimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-19-[(prop-2-en-1-yl)amino]-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate
Synonyms:
tanespimycin
Molecular Weight (g/mol):
585.7
Molecular Formula:
C31H43N3O8
InChi Key:
AYUNIORJHRXIBJ-CQFHKJEWSA-N
SMILES:
CO[C@@H]1C[C@H](C)CC2=C(NCC=C)C(=O)C=C(NC(=O)\C(C)=C/C=C/[C@@H](OC)[C@H](OC(N)=O)\C(C)=C\[C@@H](C)[C@@H]1O)C2=O
1
–
1
of
1
results
1
17-(Allylamino)-17-demethoxygeldanamycin, 99%
CAS: 75747-14-7 Molecular Formula: C31H43N3O8 Molecular Weight (g/mol): 585.698 MDL Number: MFCD04973892 InChI Key: AYUNIORJHRXIBJ-MKJFUHJFSA-N Synonym: 17-aag PubChem CID: 133688938 IUPAC Name: [(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11-dimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-21-(prop-2-enylamino)-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate SMILES: CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)NCC=C)C)OC)OC(=O)N)C)C)O)OC