accessibility menu, dialog, popup

UserName

CAS RN 7565-57-3

IUPAC Name: bromozincylium; cyclohexanide
Synonyms: bromozincylium; cyclohexanide
Molecular Weight (g/mol): 228.44
Molecular Formula: C6H11BrZn
InChi Key: PVURAUIMVICLOH-UHFFFAOYSA-M
SMILES: [Zn+]Br.C1CC[CH-]CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results