CAS RN 7488-56-4
IUPAC Name:
thione
Synonyms:
selenium disulfide
Molecular Weight (g/mol):
143.09
Molecular Formula:
S2Se
InChi Key:
JNMWHTHYDQTDQZ-UHFFFAOYSA-N
SMILES:
S=[Se]=S
1
–
2
of
2
results
1
Promotions Available
Selenium sulfide, 97%
Selenium sulfide, 97%
CAS: 7488-56-4 Molecular Formula: S2Se Molecular Weight (g/mol): 143.09 MDL Number: MFCD00011227 InChI Key: JNMWHTHYDQTDQZ-UHFFFAOYSA-N Synonym: selenium disulfide,selenium disulphide,selsun blue,selenium iv disulfide,selenium sulfide ses2,sebusan,selenex,selenix,selukos,lenium PubChem CID: 24087 IUPAC Name: thione SMILES: S=[Se]=S
2
Selenium sulfide, 94%
CAS: 7488-56-4 Molecular Formula: S2Se Molecular Weight (g/mol): 143.09 MDL Number: MFCD00011227 InChI Key: JNMWHTHYDQTDQZ-UHFFFAOYSA-N Synonym: selenium disulfide,selenium disulphide,selsun blue,selenium iv disulfide,selenium sulfide ses2,sebusan,selenex,selenix,selukos,lenium PubChem CID: 24087 IUPAC Name: thione SMILES: S=[Se]=S