accessibility menu, dialog, popup

UserName

NNNCl

CAS RN 748796-39-6

IUPAC Name: 5-(chloromethyl)-2-(1H-pyrazol-1-yl)pyridine
Synonyms: 5-(chloromethyl)-2-(pyrazol-1-yl)pyridine
Molecular Weight (g/mol): 193.63
Molecular Formula: C9H8ClN3
InChi Key: QVDLCABKNWXXQF-UHFFFAOYSA-N
SMILES: ClCC1=CN=C(C=C1)N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results