accessibility menu, dialog, popup

UserName

CAS RN 745783-83-9

IUPAC Name: 1-(1-isocyanatoethyl)-4-methoxybenzene
Synonyms: 1-(1-isocyanatoethyl)-4-methoxybenzene
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: WTNKKJUXEYSETC-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(C)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results