accessibility menu, dialog, popup

UserName

CH3NHOCH3CH3HO(E)

CAS RN 7431-25-6

IUPAC Name: (3E)-3-(hydroxyimino)-2-methylbutan-2-ol
Synonyms: (3E)-3-(hydroxyimino)-2-methylbutan-2-ol
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: QKLLBCGVADVPKD-GQCTYLIASA-N
SMILES: C\C(=N/O)C(C)(C)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results