CAS RN 7429-37-0
IUPAC Name:
(1R,2R)-1,2-dibromocyclohexane
Synonyms:
(1R,2R)-1,2-dibromocyclohexane
Molecular Weight (g/mol):
241.95
Molecular Formula:
C6H10Br2
InChi Key:
CZNHKZKWKJNOTE-PHDIDXHHSA-N
SMILES:
Br[C@@H]1CCCC[C@H]1Br
1
–
1
of
1
results
1
(+/-)-trans-1,2-Dibromocyclohexane 95.0+%, TCI America™
CAS: 7429-37-0 Molecular Formula: C6H10Br2 Molecular Weight (g/mol): 241.95 MDL Number: MFCD00003820 InChI Key: CZNHKZKWKJNOTE-PHDIDXHHSA-N Synonym: trans-1,2-dibromocyclohexane,1r,2r-1,2-dibromocyclohexane,+/--trans-1,2-dibromocyclohexane,cyclohexane,1,2-dibromo-,trans,trans-1,2-dibromcyclohexan,1beta,2alpha-dibromocyclohexane,cyclohexane,2-dibromo-, trans,1r,2r-1,2-bis bromanyl cyclohexane PubChem CID: 98506 IUPAC Name: (1R,2R)-1,2-dibromocyclohexane SMILES: Br[C@@H]1CCCC[C@H]1Br