accessibility menu, dialog, popup

UserName

H3COOOBr

CAS RN 7425-63-0

IUPAC Name: methyl 3-bromo-2-oxopropanoate
Synonyms: methyl 3-bromo-2-oxopropanoate
Molecular Weight (g/mol): 180.99
Molecular Formula: C4H5BrO3
InChi Key: MQONVZMIFQQQHA-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results