accessibility menu, dialog, popup

UserName

CAS RN 7418-16-8

IUPAC Name: 4-[2-(4-oxocyclohexyl)propan-2-yl]cyclohexan-1-one
Synonyms: 4-[2-(4-oxocyclohexyl)propan-2-yl]cyclohexan-1-one
Molecular Weight (g/mol): 236.36
Molecular Formula: C15H24O2
InChi Key: HAWVCXABNZBPED-UHFFFAOYSA-N
SMILES: CC(C)(C1CCC(=O)CC1)C1CCC(=O)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results