CAS RN 7367-81-9
CAS RN 7367-81-9
IUPAC Name:
methyl (2E)-oct-2-enoate
Synonyms:
methyl (2E)-oct-2-enoate
Molecular Weight (g/mol):
156.23
Molecular Formula:
C9H16O2
InChi Key:
CJBQSBZJDJHMLF-BQYQJAHWSA-N
SMILES:
CCCCC\C=C\C(=O)OC
1
–
1
of
1
results
1
Methyl trans-2-Octenoate 95.0+%, TCI America™
CAS: 7367-81-9 Molecular Formula: C9H16O2 Molecular Weight (g/mol): 156.225 MDL Number: MFCD00036620 InChI Key: CJBQSBZJDJHMLF-BQYQJAHWSA-N Synonym: methyl trans-2-octenoate,methyl oct-2-enoate,methyl 2-octenoate,2-octenoic acid, methyl ester,methyl e-oct-2-enoate,unii-9en9v40j78,2-octenoic acid, methyl ester, e,2-octenoic acid, methyl ester, 2e,2-octenoic acid methyl ester,fema no. 3712 PubChem CID: 5364532 IUPAC Name: methyl (E)-oct-2-enoate SMILES: CCCCCC=CC(=O)OC