accessibility menu, dialog, popup

UserName

CAS RN 73602-61-6

IUPAC Name: trihydrogen triethylamine trifluoride
Synonyms: trihydrogen triethylamine trifluoride
Molecular Weight (g/mol): 161.21
Molecular Formula: C6H18F3N
InChi Key: IKGLACJFEHSFNN-UHFFFAOYSA-N
SMILES: [H+].[H+].[H+].[F-].[F-].[F-].CCN(CC)CC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)