CAS RN 73568-27-1
IUPAC Name:
2-chloro-6-methylquinoline-3-carbaldehyde
Synonyms:
2-chloro-6-methylquinoline-3-carbaldehyde
Molecular Weight (g/mol):
205.64
Molecular Formula:
C11H8ClNO
InChi Key:
FSLNYYZJXMGKHK-UHFFFAOYSA-N
SMILES:
CC1=CC=C2N=C(Cl)C(C=O)=CC2=C1
1
–
1
of
1
results
1
2-Chloro-6-methylquinoline-3-carboxaldehyde 96.0+%, TCI America™
CAS: 73568-27-1 Molecular Formula: C11H8ClNO Molecular Weight (g/mol): 205.64 MDL Number: MFCD01912773 InChI Key: FSLNYYZJXMGKHK-UHFFFAOYSA-N Synonym: 2-Chloro-3-formyl-6-methylquinoline PubChem CID: 689081 IUPAC Name: 2-chloro-6-methylquinoline-3-carbaldehyde SMILES: CC1=CC=C2N=C(Cl)C(C=O)=CC2=C1