accessibility menu, dialog, popup

UserName

CH3NOBr

CAS RN 733744-98-4

IUPAC Name: 4-[(4-bromophenyl)(4-methylphenyl)amino]benzaldehyde
Synonyms: 4-[(4-bromophenyl)(4-methylphenyl)amino]benzaldehyde
Molecular Weight (g/mol): 366.26
Molecular Formula: C20H16BrNO
InChi Key: SDKPEZBGWABWKD-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results