accessibility menu, dialog, popup

UserName

CAS RN 7312-14-3

IUPAC Name: 5-bromo-1-benzothiophene-2-carbonyl chloride
Synonyms: 5-bromo-1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 275.54
Molecular Formula: C9H4BrClOS
InChi Key: LRYLQGGHNAOVLT-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC2=CC(Br)=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results