accessibility menu, dialog, popup

UserName

CAS RN 73025-93-1

H3CNS

CAS RN 73025-93-1

IUPAC Name: 10-hexyl-10H-phenothiazine
Synonyms: 10-hexylphenothiazine
Molecular Weight (g/mol): 283.43
Molecular Formula: C18H21NS
InChi Key: VRJVOTTZESKWPD-UHFFFAOYSA-N
SMILES: CCCCCCN1C2=CC=CC=C2SC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results