CAS RN 7298-89-7
IUPAC Name:
2-chloro-5,5-dimethylcyclohexane-1,3-dione
Synonyms:
2-chloro-5,5-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol):
174.62
Molecular Formula:
C8H11ClO2
InChi Key:
VOBIHUAWDXUCPH-UHFFFAOYSA-N
SMILES:
CC1(C)CC(=O)C(Cl)C(=O)C1
1
–
1
of
1
results
1
Promotions Available
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione, 98%
CAS: 7298-89-7 Molecular Formula: C8H11ClO2 Molecular Weight (g/mol): 174.624 MDL Number: MFCD00010502 InChI Key: VOBIHUAWDXUCPH-UHFFFAOYSA-N Synonym: chlorodimedone,2-chlorodimedone,monochlorodimedone,2-chloro-5,5-dimethyl-1,3-cyclohexanedione,1,1-dimethyl-4-chloro-3,5-cyclohexanedione,1,3-cyclohexanedione, 2-chloro-5,5-dimethyl,monochlorodimedon PubChem CID: 122278 IUPAC Name: 2-chloro-5,5-dimethylcyclohexane-1,3-dione SMILES: CC1(CC(=O)C(C(=O)C1)Cl)C