accessibility menu, dialog, popup

UserName

CAS RN 7296-20-0

IUPAC Name: hydrogen 2,9-dimethyl-1,10-phenanthroline chloride
Synonyms: hydrogen neocuproine chloride
Molecular Weight (g/mol): 244.72
Molecular Formula: C14H13ClN2
InChi Key: QUGBNSJLQDNNPK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C2C=CC3=CC=C(C)N=C3C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results