CAS RN 72411-52-0
IUPAC Name:
5-(4-fluorophenyl)-1H-pyrazol-3-amine
Synonyms:
5-(4-fluorophenyl)-1H-pyrazol-3-amine
Molecular Weight (g/mol):
177.18
Molecular Formula:
C9H8FN3
InChi Key:
QYEHDCXFXONDPV-UHFFFAOYSA-N
SMILES:
NC1=NNC(=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
5-Amino-3-(4-fluorophenyl)-1H-pyrazole, 97%
5-Amino-3-(4-fluorophenyl)-1H-pyrazole, 97%
CAS: 72411-52-0 Molecular Formula: C9H8FN3 Molecular Weight (g/mol): 177.18 MDL Number: MFCD01023677 InChI Key: QYEHDCXFXONDPV-UHFFFAOYSA-N Synonym: 3-4-fluorophenyl-1h-pyrazol-5-amine,5-4-fluorophenyl-1h-pyrazol-3-amine,5-4-fluorophenyl-2h-pyrazol-3-ylamine,5-amino-3-4-fluorophenyl-1h-pyrazole,5-amino-3-4-fluorophenyl pyrazole,1h-pyrazol-3-amine,5-4-fluorophenyl,5-4-fluorophenyl-2h-pyrazol-3-amine,3-amino-5-4-fluorophenyl-1h-pyrazole,5-4-fluoro-phenyl-2h-pyrazol-3-ylamine,3-4-fluorophenyl pyrazole-5-ylamine PubChem CID: 2759138 IUPAC Name: 5-(4-fluorophenyl)-1H-pyrazol-3-amine SMILES: NC1=NNC(=C1)C1=CC=C(F)C=C1