CAS RN 72259-10-0
CAS RN 72259-10-0
IUPAC Nor:
copper(2+) bis(tetrafluoroboranuide) hexahydrate
Synonymes:
copper(2+) hexahydrate ditetrafluoroborate
Poids moléculaire (g/mol):
345.24
Formule moléculaire:
B2CuF8H12O6
InChi Key:
FBYIXFBIARQMJJ-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F
1
–
1
de
1
résultats
1
Copper(II) tetrafluoroborate hexahydrate, 98%
CAS: 72259-10-0 Formule moléculaire: B2CuF8H12O6 Poids moléculaire (g/mol): 345.24 Numéro MDL: MFCD05863735 Clé InChI: FBYIXFBIARQMJJ-UHFFFAOYSA-N Synonyme: copper ii tetrafluoroborate hexahydrate,copper 2+ hexahydrate ditetrafluoroborate,copper ditetrafluoroborate hexahydrate,copper 2+ tetrafluoroborate-water 1/2/6,copper 2+ ion hexahydrate ditetrafluoroborate,borate 1-,tetrafluoro-, copper 2+ 2:1 , hexahydrate 9ci CID PubChem: 15774553 Nom IUPAC: copper(2+) bis(tetrafluoroboranuide) hexahydrate SMILES: O.O.O.O.O.O.[Cu++].F[B-](F)(F)F.F[B-](F)(F)F