accessibility menu, dialog, popup

UserName

CAS RN 7203-96-5

IUPAC Name: azepane-2-thione
Synonyms: 2-azacycloheptanthione
Molecular Weight (g/mol): 129.22
Molecular Formula: C6H11NS
InChi Key: APLHDUWNMGJBFD-UHFFFAOYSA-N
SMILES: S=C1CCCCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results