accessibility menu, dialog, popup

UserName

CAS RN 71987-67-2

IUPAC Name: 2-(5-fluoro-2-methyl-1H-indol-3-yl)acetic acid
Synonyms: (5-fluoro-2-methyl-1H-indol-3-yl)acetic acid
Molecular Weight (g/mol): 207.2
Molecular Formula: C11H10FNO2
InChi Key: VJZAMNBVIWOUJR-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C2=CC(F)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results