accessibility menu, dialog, popup

UserName

CAS RN 719-80-2

H3COP

CAS RN 719-80-2

IUPAC Name: ethyl diphenylphosphinite
Synonyms: ethyl diphenylphosphinite
Molecular Weight (g/mol): 230.25
Molecular Formula: C14H15OP
InChi Key: JCRCPEDXAHDCAJ-UHFFFAOYSA-N
SMILES: CCOP(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results