accessibility menu, dialog, popup

UserName

CH3ONH2H2NHCl

CAS RN 71810-97-4

IUPAC Name: 2-aminopropanamide hydrochloride
Synonyms: 2-aminopropanamide hydrochloride
Molecular Weight (g/mol): 124.57
Molecular Formula: C3H9ClN2O
InChi Key: FIAINKIUSZGVGX-UHFFFAOYNA-N
SMILES: Cl.CC(N)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results