accessibility menu, dialog, popup

UserName

CAS RN 71789-10-1

IUPAC Name: trimethyl(penta-1,4-diyn-1-yl)silane
Synonyms: trimethyl(penta-1,4-diyn-1-yl)silane
Molecular Weight (g/mol): 136.27
Molecular Formula: C8H12Si
InChi Key: JKYMNPFNLMDSFG-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCC#C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results