man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 7170-36-7

CH3OONOHO

CAS RN 7170-36-7

IUPAC Name: 6-(methoxycarbonyl)pyridine-2-carboxylic acid
Synonyms: 6-(methoxycarbonyl)pyridine-2-carboxylic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: VWIOMFMPIVMLIR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(=N1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results